Skip to Content
Merck
All Photos(1)

Key Documents

378615

Sigma-Aldrich

3,3-Diethoxypropionitrile

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2CHCH2CN
CAS Number:
Molecular Weight:
143.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
95%

Assay

95%

form

liquid

refractive index

n20/D 1.415 (lit.)

bp

91-93 °C/11 mmHg (lit.)

density

0.954 g/mL at 25 °C (lit.)

functional group

ether
nitrile

SMILES string

CCOC(CC#N)OCC

InChI

1S/C7H13NO2/c1-3-9-7(5-6-8)10-4-2/h7H,3-5H2,1-2H3

InChI key

WBOXEOCWOCJQNK-UHFFFAOYSA-N

General description

3,3-Diethoxypropionitrile is a cyanide derivative. It undergoes reaction with urea in refluxing butanolic sodium butoxide to afford 4-amino-2(1H)-pyrimidinone and cytosine. Its physical properties (density, refractive index, boiling point and melting point) have been reported.

Application

3,3-Diethoxypropionitrile may be used in the synthesis of 2-(2,2-diethoxy-ethyl)-pyridine, via [2+2+2]-cycloaddition reaction. It may be used in the synthesis of cyanomalondialdehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation, Structure, and Reactions of Halomalondialehdes.
Reichardt C and Halbritter K.
Angewandte Chemie (International Edition in English), 14(2), 86-94 (1975)
Brown DJ.
The Chemistry of Heterocyclic Compounds, 216-216 (2009)
Synthesis of isoxazole.
Cai D, et al.
Chemical Reagents, 1, 027-027 (2010)
Barbara Heller et al.
The Journal of organic chemistry, 67(13), 4414-4422 (2002-06-22)
The photocatalyzed [2 + 2 + 2]-cycloaddition of nitriles with 2 equiv of acetylene to 2-pyridines can be carried out under mild conditions and represents a valuable extension to common synthetical methods. For the ideal wavelength range (350-500 nm), lamps
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 164-164 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service