Skip to Content
Merck
All Photos(1)

Documents

378283

Sigma-Aldrich

Benzyltrimethylsilane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2Si(CH3)3
CAS Number:
Molecular Weight:
164.32
Beilstein:
1928113
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.493 (lit.)

bp

187-190 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)Cc1ccccc1

InChI

1S/C10H16Si/c1-11(2,3)9-10-7-5-4-6-8-10/h4-8H,9H2,1-3H3

InChI key

MRIWRLGWLMRJIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bulletin de la Societe Chimique De France, 90, 11-11 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service