Skip to Content
Merck
All Photos(1)

Documents

376833

Sigma-Aldrich

2-Chloro-5-(trifluoromethyl)benzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)CO2H
CAS Number:
Molecular Weight:
224.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

93-96 °C (lit.)

SMILES string

OC(=O)c1cc(ccc1Cl)C(F)(F)F

InChI

1S/C8H4ClF3O2/c9-6-2-1-4(8(10,11)12)3-5(6)7(13)14/h1-3H,(H,13,14)

InChI key

WLXRKCGYQAKHSJ-UHFFFAOYSA-N

General description

2-Chloro-5-(trifluoromethyl)benzoic acid is a carboxylic acid building block.

Application

2-Chloro-5-(trifluoromethyl)benzoic acid may be used as surrogate standard to investigate the pesticide exposure and home contamination in urine and hand wipe samples collected from farmers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian D Curwin et al.
Journal of exposure analysis and environmental epidemiology, 15(6), 500-508 (2005-04-21)
In the spring and summer of 2001, as part of a larger study investigating farm family pesticide exposure and home contamination in Iowa, urine and hand wipe samples were collected from 24 male farmers and 23 male nonfarmer controls. On

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service