Skip to Content
Merck
All Photos(2)

Key Documents

373699

Sigma-Aldrich

2,3-O-Isopropylidene-D-ribonic γ-lactone

99%

Synonym(s):

2,3-O-Isopropylidene-D-ribono-1,4-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O5
CAS Number:
Molecular Weight:
188.18
Beilstein:
10090
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −69°, c = 2 in pyridine

mp

135-138 °C (lit.)

SMILES string

CC1(C)O[C@@H]2[C@@H](CO)OC(=O)[C@H]2O1

InChI

1S/C8H12O5/c1-8(2)12-5-4(3-9)11-7(10)6(5)13-8/h4-6,9H,3H2,1-2H3/t4-,5-,6-/m1/s1

InChI key

NHHKFJCWLPPNCN-HSUXUTPPSA-N

Application

Important raw material for the synthesis of C-nucleosides, prostanoids, and GABA analogs via homochiral 2-piperidones. Starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi,Z. et al.
Tetrahedron, 48, 349-349 (1992)
Marquez, V.E. et al.
Nucl. Acid Chem., 4, 27-27 (1991)
Nucl. Acid Chem., 4, 27-27 (1991)
Tetrahedron Letters, 29, 5521-5521 (1988)
Herdeis, C. et al.
Naunyn-Schmiedeberg'S Archives of Pharmacology, 324, 269-269 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service