Skip to Content
Merck
All Photos(1)

Key Documents

370118

Sigma-Aldrich

4-(trans-4-Pentylcyclohexyl)benzonitrile

99%, liquid crystal (nematic)

Synonym(s):

4-(4-Pentylcyclohexyl)benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C6H10C6H4CN
CAS Number:
Molecular Weight:
255.40
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

form

liquid crystal (nematic)

mp

30-55 °C (lit.)

SMILES string

CCCCC[C@@H]1CC[C@H](CC1)c2ccc(cc2)C#N

InChI

1S/C18H25N/c1-2-3-4-5-15-6-10-17(11-7-15)18-12-8-16(14-19)9-13-18/h8-9,12-13,15,17H,2-7,10-11H2,1H3/t15-,17-

InChI key

FURZYCFZFBYJBT-JCNLHEQBSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chiral nematic liquid crystal (N*-LC) facilitates asymmetric acetylene polymerization, yielding helical chains and fibrils in polyacetylene film.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service