Skip to Content
Merck
All Photos(1)

Key Documents

367249

Sigma-Aldrich

(1R,2S)-trans-2-Phenyl-1-cyclohexanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H10OH
CAS Number:
Molecular Weight:
176.25
Beilstein:
2047602
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D −58°, c = 10 in methanol

optical purity

ee: 99% (GLC)

bp

276-281 °C (lit.)

mp

63-66 °C (lit.)

SMILES string

O[C@@H]1CCCC[C@H]1c2ccccc2

InChI

1S/C12H16O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-3,6-7,11-13H,4-5,8-9H2/t11-,12+/m0/s1

InChI key

AAIBYZBZXNWTPP-NWDGAFQWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare chiral isoprenyl[1] and vinyl[2] ethers and chiral pyruvates.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The resolution of racemic hydroperoxides: the preparation of optically pure hydroperoxide natural products.
P Dussault et al.
Journal of the American Chemical Society, 110(18), 6276-6277 (1988-08-01)
The Journal of Organic Chemistry, 54, 2258-2258 (1989)
Synthesis, 836-836 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service