Skip to Content
Merck
All Photos(1)

Key Documents

362956

Sigma-Aldrich

Calcium bis(2,2,6,6-tetramethyl-3,5-heptanedionate)

Synonym(s):

Ca(TMHD)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OCC(CH3)3CHCOC(CH3)3)2
CAS Number:
Molecular Weight:
406.61
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Ca, 8.7-11.0% EDTA titration

reaction suitability

core: calcium

mp

221-224 °C (lit.)

SMILES string

CC(C)(C)C(=O)\C=C(/O[Ca]O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/2C11H20O2.Ca/c2*1-10(2,3)8(12)7-9(13)11(4,5)6;/h2*7,12H,1-6H3;/q;;+2/p-2/b2*8-7-;

InChI key

DOOFPPIHJGRIGW-ATMONBRVSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used in the preparation of thin-film superconductors.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 23, 54-54 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service