Skip to Content
Merck
All Photos(1)

Key Documents

36070

Sigma-Aldrich

2,6-Dichloro-4-nitrophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2NO3
CAS Number:
Molecular Weight:
208.00
Beilstein:
1245045
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.5% (HPLC)
98%

form

solid

mp

123-126 °C (dec.)

solubility

methanol: soluble 1 g/10 mL, clear, slightly yellow to deep greenish-yellow

SMILES string

Oc1c(Cl)cc(cc1Cl)[N+]([O-])=O

InChI

1S/C6H3Cl2NO3/c7-4-1-3(9(11)12)2-5(8)6(4)10/h1-2,10H

InChI key

PXSGFTWBZNPNIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingxing Diao et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(4), 774-781 (2014-01-29)
3-n-Butylphthalide (NBP) [(±)-3-butyl-1(3H)-isobenzofuranone] is an anti-cerebral-ischemia drug. Moderate hepatotoxicity has been observed in clinical applications. One of the major metabolites, 3-N-acetylcysteine-NBP, has been detected in human urine, indicating the formation of a reactive metabolite. We elucidated the formation mechanism of
H Koster et al.
Biochemical pharmacology, 31(10), 1919-1924 (1982-05-15)
The pharmacokinetics of 2,6-dichloro-4-nitrophenol (DCNP) have been studied in the rat. Upon i.v. injection the plasma decay curve of DCNP showed a rapid distribution phase. After 30 min the plasma concentration reached a value that was constant for at least
Y Nakagawa et al.
Toxicology, 156(1), 27-36 (2001-02-13)
The metabolism and cytotoxicity of benzophenone and estrogenic activity of its metabolites have been studied in freshly isolated rat hepatocytes and cultured MCF-7 human breast cancer cells, respectively. The incubation of hepatocytes with benzophenone (0.25-1.0 mM) elicited a concentration- and
K O Wong et al.
Xenobiotica; the fate of foreign compounds in biological systems, 26(1), 17-26 (1996-01-01)
1. Amine N-sulphotransferase (NST) activity with desipramine (DMI) as substrate was assayed in vitro in various areas of the rat brain. Biosynthesis of 3'-phosphoadenosine-5'-phospho35sulphate (PAPS) from sodium 35sulphate and ATP was also measured by coupling it to the sulphation of
Andreas Wild et al.
Chemical communications (Cambridge, England), 48(7), 964-966 (2011-12-14)
Water-based Zn(II) bisterpyridine systems were used as fluorometric sensors for the detection of the nerve gas G mimics DMMP, DCP and DCNP. Analyte concentrations in the range of 10(-7) to 10(-6) M are detectable in solution. The utilization of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service