Skip to Content
Merck
All Photos(1)

Documents

358878

Sigma-Aldrich

N,N′-Ethylenebis(acrylamide)

technical grade, ≥90% (TLC)

Synonym(s):

N,N′-Dimethylenebis(acrylamide), N,N′-Bisacryloyl-1,2-diaminoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCONHCH2-)2
CAS Number:
Molecular Weight:
168.19
Beilstein:
1771570
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

≥90% (TLC)

mp

138-140 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC(=O)NCCNC(=O)C=C

InChI

1S/C8H12N2O2/c1-3-7(11)9-5-6-10-8(12)4-2/h3-4H,1-2,5-6H2,(H,9,11)(H,10,12)

InChI key

AYGYHGXUJBFUJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiran Reddy Kanubaddi et al.
Nanomaterials (Basel, Switzerland), 11(2) (2021-01-31)
In this study, a methodology utilizing peptide conformational imprints (PCIs) as a tool to specifically immobilize porcine pancreatic alpha-trypsin (PPT) at a targeted position is demonstrated. Owing to the fabrication of segment-mediated PCIs on the magnetic particles (PCIMPs), elegant cavities
Giuseppe Cirillo et al.
Materials science & engineering. C, Materials for biological applications, 48, 499-510 (2015-01-13)
Thermo-responsive hydrogel films, synthesized by UV-initiated radical polymerization, are proposed as delivery devices for non-steroidal anti-inflammatory drugs (Diclofenac sodium and Naproxen). N-isopropylacrylamide and N,N'-ethylenebisacrylamide were chosen as thermo-sensitive monomer and crosslinker, respectively. Infrared spectroscopy was used to assess the incorporation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service