Skip to Content
Merck
All Photos(1)

Key Documents

349658

Sigma-Aldrich

N-(2-Chloroethyl)phthalimide

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8ClNO2
CAS Number:
Molecular Weight:
209.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

82-84 °C (lit.)

functional group

chloro
imide

SMILES string

ClCCN1C(=O)c2ccccc2C1=O

InChI

1S/C10H8ClNO2/c11-5-6-12-9(13)7-3-1-2-4-8(7)10(12)14/h1-4H,5-6H2

InChI key

ZPHGARLYQHMNTB-UHFFFAOYSA-N

General description

N-(2-Chloroethyl)phthalimide can be prepared by the reaction of potassium phthalimide and 2-chloroethyl tosylate.

Application

N-(2-Chloroethyl)phthalimide may be used in the preparation of 1-[(dialkylamino)alkyl]-1,8-napthyridin-2(1H)ones and were evaluated for the inhibition of gastric acid secretion invivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Gabriel synthesis of primary amines.
Gibson MS and Bradshaw RW.
Angewandte Chemie (International Edition in English), 7(12), 919-930 (1968)
Inhibition of gastric acid secretion by 1,8-naphthyridin-2(1H)-ones.
W A Bolhofer et al.
Journal of medicinal chemistry, 22(3), 301-306 (1979-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service