Skip to Content
Merck
All Photos(1)

Key Documents

348899

Sigma-Aldrich

2,6-Dichloro-3-nitrobenzoic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)CO2H
CAS Number:
Molecular Weight:
236.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

152-158 °C (lit.)

SMILES string

OC(=O)c1c(Cl)ccc(c1Cl)[N+]([O-])=O

InChI

1S/C7H3Cl2NO4/c8-3-1-2-4(10(13)14)6(9)5(3)7(11)12/h1-2H,(H,11,12)

InChI key

PCIHIUCCINHORT-UHFFFAOYSA-N

General description

FT-IR and Raman spectra of 2, 6-dichloro-3-nitrobenzoic acid (DCNBA) have been investigated.[1]

Application

2,6-Dichloro-3-nitrobenzoic acid may be used in the preparation of 9-chloro-2-methoxy-6-nitro-5,10-dihydrobenzo[b][1,5]naphthyridin-10-one, required for the synthesis of potential DNA-binding antitumor agents.[2] It may be used in the preparation of 2-(aminoalkyl)-5-nitropyrazolo [3,4,5-kl] acridines.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D B Capps et al.
Journal of medicinal chemistry, 35(26), 4770-4778 (1992-12-25)
2-(Aminoalkyl)-5-nitropyrazolo[3,4,5-kl]acridines were prepared from substituted anilines via the 1-chloro-4-nitroacridones followed by condensation with [(alkylamino)alkyl]hydrazines. Impressive activity was demonstrated for the 9-hydroxy, 9-alkoxy, and 9-acyloxy analogs in vitro on a L1210 leukemia line and in vivo against the P388 leukemia. Advanced
Amelia Magnano et al.
Bioorganic & medicinal chemistry, 12(22), 5941-5947 (2004-10-23)
A series of potential DNA-binding antitumor agents, 2-[omega-(alkylamino)alkyl]-9-methoxy-5-nitro-2,6-dihydroindazolo[4,3-bc][1,5]naphthyridines (2a-f), 10-aza derivatives of PZA, has been prepared by condensation of 9-chloro-2-methoxy-6-nitro-5,10-dihydrobenzo[b][1,5]naphthyridin-10-one (6) with the appropriate (omega-aminoalkyl)hydrazine in tetrahydrofuran/methanol. Compound 6 was obtained by heating at 100 degrees C in H(2)SO(4)5, yielded
V Balachandran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 110, 130-140 (2013-04-09)
The FT-IR and Raman spectra of 2, 6-dichloro-3-nitrobenzoic acid (DCNBA) have been recorded and analyzed. The equilibrium geometry, various bonding and harmonic vibrational wavenumbers have been calculated with the help of density functional theory (DFT/B3LYP/cc-pvdz/6-311++G(d,p)) method. The optimized geometrical parameters

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service