Skip to Content
Merck
All Photos(1)

Key Documents

345660

Sigma-Aldrich

1,2-Dichloro-4-fluoro-5-nitrobenzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(F)NO2
CAS Number:
Molecular Weight:
209.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.575 (lit.)

bp

247 °C (lit.)

mp

17 °C (lit.)

density

1.596 g/mL at 25 °C (lit.)

functional group

chloro
fluoro
nitro

SMILES string

[O-][N+](=O)c1cc(Cl)c(Cl)cc1F

InChI

1S/C6H2Cl2FNO2/c7-3-1-5(9)6(10(11)12)2-4(3)8/h1-2H

InChI key

FXOCDIKCKFOUDE-UHFFFAOYSA-N

General description

Proton-detected local field (PDLF) spectra of 1,2-dichloro-4-fluoro-5-nitrobenzene in nematic solvents has been studied.

Application

1,2-Dichloro-4-fluoro-5-nitrobenzene may be used in the combinatorial solid-phase synthesis of bis-heterocyclic compounds. It was employed as solute to investigate the possibility of selective excitation in coupled multispin systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miroslav Soural et al.
Journal of combinatorial chemistry, 10(6), 923-933 (2008-09-25)
Combinatorial solid-phase synthesis of bis-heterocyclic compounds, characterized by the presence of two heterocyclic cores connected by a spacer of variable length/structure, provided structurally heterogeneous libraries with skeletal diversity. Both heterocyclic rings were assembled on resin in a combinatorial fashion.
Selective excitation in dipole coupled systems.
Walls JD, et al.
Chemical Physics Letters, 357(3), 241-248 (2002)
Measurement of dipolar couplings in partially oriented molecules by local field NMR spectroscopy with low-power decoupling.
Marjanska M, et al.
Journal of Magnetic Resonance, 158(1), 52-59 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service