Skip to Content
Merck
All Photos(1)

Key Documents

345067

Sigma-Aldrich

2-Fluoro-4-nitrophenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)OH
CAS Number:
Molecular Weight:
157.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Assay

99%

mp

120-122 °C (lit.)

SMILES string

Oc1ccc(cc1F)[N+]([O-])=O

InChI

1S/C6H4FNO3/c7-5-3-4(8(10)11)1-2-6(5)9/h1-3,9H

InChI key

ORPHLVJBJOCHBR-UHFFFAOYSA-N

General description

2-Fluoro-4-nitrophenol is a derivative of m-fluoronitrobenzene. Crystal structure of complex of α-cyclodextrin with 2-fluoro-4-nitrophenol has been studied by the X-ray diffraction technique.[1]

Application

2-Fluoro-4-nitrophenol may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koya Yamaguchi et al.
Journal of the American Chemical Society, 133(36), 14208-14211 (2011-08-20)
Monoamine oxidase (MAO) A is a flavoenzyme that catalyzes the oxidation of biologically important monoamines and is thought to be associated with psychiatric disorders. Here, we report a strategy for rationally designing a (19)F magnetic resonance imaging probe for the
M Shibakami et al.
Carbohydrate research, 260(2), 169-179 (1994-07-16)
The crystal structure of a complex of alpha-cyclodextrin (alpha-CD) with 2-fluoro-4-nitrophenol.3H2O has been determined by the X-ray diffraction technique. The complex crystallizes in space group P2(1)2(1)2(1) with cell dimensions: a = 13.431(3), b = 15.299(4), c = 24.780(5) A. The
Li Liu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 21(9), 2014-2019 (2007-03-14)
Gene therapy shows promise for treating prostate cancer and has been evaluated in several clinical trials. A major challenge that remains is to establish a method for verifying transgene activity in situ. The lacZ gene encoding beta-galactosidase historically has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service