Skip to Content
Merck
All Photos(1)

Documents

344311

Sigma-Aldrich

2,5-Dimethyl-2,4-hexadiene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH=C(CH3)2
CAS Number:
Molecular Weight:
110.20
Beilstein:
1733342
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.8 (vs air)

vapor pressure

26.9 mmHg ( 37.7 °C)
7.2 mmHg ( 20 °C)

Assay

96%

form

liquid

autoignition temp.

559 °F

refractive index

n20/D 1.476 (lit.)

bp

132-134 °C (lit.)

mp

11-14 °C (lit.)

density

0.773 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C/C=C(\C)C

InChI

1S/C8H14/c1-7(2)5-6-8(3)4/h5-6H,1-4H3

InChI key

DZPCYXCBXGQBRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dimethyl-2,4-hexadiene is an electron-rich alkene. It induces photodechlorination of 9,10-dichloroanthracene and its mechanism has been investigated. Asymmetric cyclopropanation of 2,5-dimethyl-2,4-hexadiene with tert-butyl diazoacetate catalyzed by the new copper-bisoxazoline complex has been reported. 2,5-Dimethyl-2,4-hexadiene has been investigated as a singlet oxygen acceptor in various solvents. Mechanism of addition reaction of aromatic and aliphatic thiols with 2,5-dimethyl-2,4-hexadiene has been investigated.

Application

2,5-Dimethyl-2,4-hexadiene may be used in the synthesis of (+)-trans-(1R,3R)-chrysanthemic acid methyl ester.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Katsuhiro Suenobu et al.
Journal of the American Chemical Society, 126(23), 7271-7280 (2004-06-10)
The reaction pathway and the mechanism of asymmetric induction in the synthesis of (+)-trans-(1R,3R)-chrysanthemic acid methyl ester from methyl diazoacetate and 2,5-dimethyl-2,4-hexadiene in the presence of a C(1)-chiral salicylaldimine Cu(I) complex has been probed with the aid of hybrid density
Makoto Itagaki et al.
The Journal of organic chemistry, 70(8), 3292-3295 (2005-04-13)
Some new bisoxazoline ligands with an aryl group at the 4-position and gem-dimethyl groups at the 5-position on the oxazoline ring were prepared from arylglycines. Remarkable enhancement of the trans-selectivity (trans/cis = 87/13) and the enantioselectivity (96% ee for the
Organic sulfur compounds. VII. Some addition and co-oxidation reactions of thiols with 2, 5-dimethyl-2, 4-hexadiene.
Oswald AA, et al.
The Journal of Organic Chemistry, 27(7), 2439-2448 (1962)
Interactions of singlet oxygen with 2, 5-dimethyl-2, 4-hexadiene in polar ano non-polar solvents evidence for a vinylog ene-reaction.
Gollnick K and Griesbeck A.
Tetrahedron, 40(17), 3235-3250 (1984)
J Saltiel et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 8(6), 856-867 (2009-06-06)
Photochemical formation of 9-chloroanthracene (MCA) from 9,10-dichloroanthracene (DCA) is observed in the presence of 2,5-dimethyl-2,4-hexadiene (DMH) in acetonitrile (AN). The mechanism of the reaction was investigated using kinetics, deuterium labeling, and quenching techniques. Contrary to conclusions in a recent publication

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service