Skip to Content
Merck
All Photos(1)

Key Documents

340448

Sigma-Aldrich

Dipotassium deuterium phosphate

98 atom % D

Synonym(s):

Phosphoric acid-d dipotassium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$3,827.00

R$3,827.00


Estimated to ship onApril 16, 2025


Request more information

Select a Size

Change View
10 G
R$3,827.00

About This Item

Linear Formula:
K2DPO4
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

R$3,827.00


Estimated to ship onApril 16, 2025


Request more information

isotopic purity

98 atom % D

Quality Level

form

powder

technique(s)

NMR: suitable

mass shift

M+1

SMILES string

[K+].[K+].[2H]OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2/i/hD

InChI key

ZPWVASYFFYYZEW-DYCDLGHISA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaqi Fu et al.
Biotechnology progress, 37(1), e3075-e3075 (2020-09-02)
Dehaloperoxidase (DHP) catalyzes detoxifying halophenols. It is a heme-containing enzyme using H2 O2 as the oxidant. Heme bleaching from the active site is of great concern. In addition, the interference of DHP by H2 O2 leads to the inactivation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service