Skip to Content
Merck
All Photos(1)

Documents

340146

Sigma-Aldrich

Glycidyl 2-methylphenyl ether

technical grade, 90%

Synonym(s):

(2-Methylphenoxymethyl)oxirane, 1,2-Epoxy-3-(2-methylphenoxy)propane, 1-(2,3-Epoxypropoxy)-2-methylbenzene, 1-(2-Methylphenoxy)-2,3-epoxypropane, 2-Methylphenyl glycidyl ether, 2-[(2-Methylphenoxy)methyl]oxirane, o -Cresol glycidyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

vapor density

>1 (vs air)

Assay

90%

form

liquid

refractive index

n20/D 1.529 (lit.)

bp

109-111 °C/4 mmHg (lit.)

density

1.079 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1OCC2CO2

InChI

1S/C10H12O2/c1-8-4-2-3-5-10(8)12-7-9-6-11-9/h2-5,9H,6-7H2,1H3

InChI key

KFUSXMDYOPXKKT-UHFFFAOYSA-N

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service