Skip to Content
Merck
All Photos(1)

Key Documents

339156

Sigma-Aldrich

Barium thiocyanate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(SCN)2 · xH2O
CAS Number:
Molecular Weight:
253.49 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

SMILES string

O.N#CS[Ba]SC#N

InChI

1S/2CHNS.Ba.H2O/c2*2-1-3;;/h2*3H;;1H2/q;;+2;/p-2

InChI key

RPIMIEHGCRZMML-UHFFFAOYSA-L

General description

The phosphorescence emission spectra and lifetime of the thiocyanate salt of barium has been measured.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lower-Energy Electronic States of HNCS, NCS-, and Thiocyanate Salts.
McDonald JR, et al.
J. Chem. Phys. , 51(5), 1723-1731 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service