Skip to Content
Merck
All Photos(1)

Key Documents

335045

Sigma-Aldrich

Ferroceneacetic acid

98%

Synonym(s):

FAA, NSC 128978

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12FeO2
CAS Number:
Molecular Weight:
244.07
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

core: iron
reagent type: catalyst

mp

158-160 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.OC(=O)C[C]2[CH][CH][CH][CH]2

InChI

1S/C7H7O2.C5H5.Fe/c8-7(9)5-6-3-1-2-4-6;1-2-4-5-3-1;/h1-4H,5H2,(H,8,9);1-5H;

InChI key

DLJGQTUEZOSNKQ-UHFFFAOYSA-N

Application

Uses:
  • Preparation of NADH oxidase-based NADH biosensors
  • Catalyst in hydroxylation and polymerization reactions
  • Reducing agent for reduction of Escherichia coli ribonucleotide reductase subunit R2
  • Antitumor agent via generation of active oxygen species, which induce oxidative DNA damage in cancer cells
  • Catalyst for oxidation of ascorbic acid

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qasim Ali et al.
Biomolecules, 10(9) (2020-08-23)
Different techniques are being employed to reduce the adverse effects of water stress on seed yield and quality of crop plants. The current study aimed to improve the water stress tolerance of field-grown sunflower by foliar-supplied ecofriendly iron-chelated aspartate (Fe-Asp)
Caihuan Tian et al.
Nature communications, 10(1), 141-141 (2019-01-13)
Gene regulatory networks control development via domain-specific gene expression. In seed plants, self-renewing stem cells located in the shoot apical meristem (SAM) produce leaves from the SAM peripheral zone. After initiation, leaves develop polarity patterns to form a planar shape.
Antonia Yarur et al.
Plant reproduction, 29(4), 311-322 (2016-11-24)
FT gene is expressed in leaves and buds and is involved in floral meristem determination and bud development in sweet cherry. In woody fruit perennial trees, floral determination, dormancy and bloom, depends on perception of different environmental and endogenous cues
Francesca Valerio et al.
Food chemistry, 221, 582-589 (2016-12-17)
A Lactobacillus plantarum fermentation product (Bio21B), obtained after strain growth (14h) in a wheat flour-based medium, was applied in the bread-making process as taste enhancer, in order to obtain a yeast-leavened bread with reduced salt content (20% and 50%) with
Takashi Akagi et al.
The Plant cell, 30(4), 780-795 (2018-04-08)
Dioecy, the presence of male and female flowers on distinct individuals, has evolved independently in multiple plant lineages, and the genes involved in this differential development are just starting to be uncovered in a few species. Here, we used genomic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service