Skip to Content
Merck
All Photos(1)

Key Documents

332259

Sigma-Aldrich

Ethyl trans-β-methylcinnamate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,124.00

R$1,124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$1,124.00

About This Item

Linear Formula:
C6H5C(CH3)=CHCO2C2H5
CAS Number:
Molecular Weight:
190.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,124.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.546 (lit.)

bp

142-144 °C/15 mmHg (lit.)

density

1.042 g/mL at 25 °C (lit.)

functional group

ester
phenyl

SMILES string

[H]\C(C(=O)OCC)=C(\C)c1ccccc1

InChI

1S/C12H14O2/c1-3-14-12(13)9-10(2)11-7-5-4-6-8-11/h4-9H,3H2,1-2H3/b10-9+

InChI key

BSXHSWOMMFBMLL-MDZDMXLPSA-N

Application

Ethyl trans-β-methylcinnamate was used in diastereoselective synthesis of ethyl (E)-3-methyl-3-phenylglycidate (strawberry flavoring agent).[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diastereoselective Synthesis of a Strawberry Flavoring Agent by Epoxidation of Ethyl trand-beta-Methylcinnamate.
Gayle JP, et al.
Journal of Chemical Education, 79(1), 96-96 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service