Skip to Content
Merck
All Photos(1)

Key Documents

332178

Sigma-Aldrich

DL-tert-Leucine

98%

Synonym(s):

(±)-2-Amino-3,3-dimethylbutyric acid, DL-α-tert-Butylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH(NH2)COOH
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721823
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)C(N)C(O)=O

InChI

1S/C6H13NO2/c1-6(2,3)4(7)5(8)9/h4H,7H2,1-3H3,(H,8,9)

InChI key

NPDBDJFLKKQMCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina F Zhurko et al.
Molecules (Basel, Switzerland), 25(4) (2020-02-23)
Nitroxides are broadly used as molecular probes and labels in biophysics, structural biology, and biomedical research. Resistance of a nitroxide group bearing an unpaired electron to chemical reduction with low-molecular-weight antioxidants and enzymatic systems is of critical importance for these
V W Cornish et al.
Biochemistry, 33(40), 12022-12031 (1994-10-11)
In order to gain greater insight into the effects of beta-branched amino acids on protein alpha-helices, hydrophobic amino acids with varying degrees of beta-branching, including the fully beta-substituted L-2-amino-3,3-dimethylbutanoic acid (ADBA), were incorporated into the protein T4 lysozyme. The unnatural
Philippe Bisel et al.
Organic & biomolecular chemistry, 6(15), 2655-2665 (2008-07-18)
The unique reactivity pattern elicited by the crowded tert-butyl group is highlighted by summarising characteristic applications. Starting from the use of this simple hydrocarbon moiety in chemical transformations, via its relevance in Nature and its implication in biosynthetic and biodegradation
Fernando Formaggio et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(8), 2395-2404 (2005-01-26)
Terminally protected homopeptides of tert-leucine, from the dimer to the hexamer, co-oligopeptides of tert-leucine in combination with alpha-aminoisobutyric acid or glycine residues up to the hexamer level, and simple dipeptides representing known scaffolds for catalysts in asymmetric organic reactions were
K Laumen et al.
Bioscience, biotechnology, and biochemistry, 65(9), 1977-1980 (2001-10-26)
Optically pure D-tert.-leucine was obtained by the enzymatic hydrolysis of (+/-)-N-acetyl-tert. leucine chloroethyl ester after about 50% conversion, this being catalyzed by a protease from Bacillus licheniformis (Alcalase), and subsequent acidic saponification of the recovered ester. Among the methyl, ethyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service