Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.543 (lit.)
bp
93-94 °C (lit.)
mp
−43 °C (lit.)
density
2.961 g/mL at 25 °C (lit.)
SMILES string
C[Hg]C
InChI
1S/2CH3.Hg/h2*1H3;
InChI key
ATZBPOVXVPIOMR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
41.0 °F - closed cup
Flash Point(C)
5 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Food chemistry, 134(4), 2345-2349 (2013-02-28)
A simple and rapid non-chromatographic method was developed to determine methylmercury (MeHg) and inorganic mercury (iHg) levels in muscles tissues of 10 freshwater fish species. The MeHg and iHg were determined by cold vapour atomic absorption spectrometry after alkaline wet
ACS chemical biology, 7(2), 411-420 (2011-10-27)
Mercury, one of the most toxic elements, exists in various chemical forms each with different toxicities and health implications. Some methylated mercury forms, one of which exists in fish and other seafood products, pose a potential threat, especially during embryonic
PloS one, 8(2), e55559-e55559 (2013-02-14)
Estrogen, a class of female sex steroids, is neuroprotective. Estrogen is synthesized in specific areas of the brain. There is a possibility that the de novo synthesized estrogen exerts protective effect in brain, although direct evidence for the neuroprotective function
Biological trace element research, 147(1-3), 156-164 (2011-11-26)
As a highly toxic environmental pollutant, methylmercury (MeHg) can cause neurotoxicity in animals and humans. Considering the antioxidant property of grape seed proanthocyanidin extracts (GSPE), this study was aimed to evaluate the effect of GSPE on MeHg-induced neurotoxicity in rats.
Archives of environmental contamination and toxicology, 61(4), 558-567 (2011-03-23)
Mercury (Hg), widely used in industry, is a great environmental health problem for humans and animals. Despite several reports regarding Hg toxicity, there is scarcity of data on its toxic manifestations on Sprague Dawley rats under realistic exposure conditions. Experimental
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service