Skip to Content
Merck
All Photos(5)

Key Documents

324612

Sigma-Aldrich

(S)-(−)-3-Chloro-1-phenyl-1-propanol

greener alternative

98%

Synonym(s):

α-(2-Chloroethyl)benzyl alcohol, (S)-(−)-α-(2-Chloroethyl)benzyl alcohol, (S)-(−)-3-Chloro-1-phenylpropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2CH(C6H5)OH
CAS Number:
Molecular Weight:
170.64
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]19/D −25°, c = 1 in chloroform

optical purity

ee: 99% (GLC)

greener alternative product score

old score: 100
new score: 62
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

58-60 °C (lit.)

functional group

chloro
hydroxyl
phenyl

greener alternative category

SMILES string

O[C@@H](CCCl)c1ccccc1

InChI

1S/C9H11ClO/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5,9,11H,6-7H2/t9-/m0/s1

InChI key

JZFUHAGLMZWKTF-VIFPVBQESA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom Economy”, "Enhanced Energy Efficiency" and “Use of Renewable Feedstock”. Click here to view its DOZN scorecard.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service