Skip to Content
Merck
All Photos(1)

Key Documents

320412

Sigma-Aldrich

Dimethyl disulfide

99%

Synonym(s):

DMDS, Methyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SSCH3
CAS Number:
Molecular Weight:
94.20
Beilstein:
1730824
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.24 (vs air)

vapor pressure

22 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

>572 °F

expl. lim.

16 %

refractive index

n20/D 1.525 (lit.)

bp

109 °C (lit.)

mp

−85 °C (lit.)

density

1.046 g/mL at 25 °C (lit.)

functional group

disulfide

SMILES string

CSSC

InChI

1S/C2H6S2/c1-3-4-2/h1-2H3

InChI key

WQOXQRCZOLPYPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Upper respiratory tract

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabrielle A Nevitt
Integrative and comparative biology, 51(5), 819-825 (2011-09-02)
Information transfer influences food-web dynamics in the marine environment, but infochemicals involved in these processes are only beginning to be understood. Dimethylsulfoniopropionate (DMSP) is produced by phytoplankton and other marine algae, and has been studied primarily in the context of
P Jäppinen et al.
Archives of toxicology, 67(2), 104-106 (1993-01-01)
Dimethyl sulfide in breath was determined by the gas chromatographic method in 14 persons exposed to organic reduced sulfur compounds in sulfate pulp mills. Dimethyl sulfide concentrations in breath (range 0.04-0.69 cm3/m3 were compared to the combined workplace concentrations of
Elise Dumont et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 12(14), 2596-2603 (2011-11-02)
Ground-state disulfide dissociation is a target of prime importance in structural biochemistry. A main difficulty consists in avoiding competition with carbon–sulfur and backbone scission pathways. In tandem mass spectrometry, such selectivity is afforded using transition elements or coinage-metal ions as
L Arellano-García et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 66(8), 1641-1646 (2012-08-22)
The aim of this paper was to evaluate the performance of biotrickling filters (BTFs) for treating low concentrations of dimethyl disulfide (DMDS), using different bacterial consortia adapted to consume reduced sulfur compounds under alkaline (pH ≈ 10) or neutral (pH
Chien-Jui Huang et al.
Pest management science, 68(9), 1306-1310 (2012-05-11)
Bacillus cereus C1L is a plant growth-promoting rhizobacterium and can elicit induced systemic resistance (ISR) in plants against necrotrophic pathogens. However, little is known about ISR elicitors produced by B. cereus C1L, and no ISR elicitor has been identified and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service