Skip to Content
Merck
All Photos(1)

Key Documents

31942

Sigma-Aldrich

(Diethylamino)sulfur trifluoride

technical, packed in PTFE bottles, ≥90% (NMR)

Synonym(s):

DAST, Diethylaminosulfur trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NSF3
CAS Number:
Molecular Weight:
161.19
Beilstein:
1849066
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (NMR)

quality

packed in PTFE bottles

bp

30-32 °C/3 mmHg (lit.)

density

1.22 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN(CC)S(F)(F)F

InChI

1S/C4H10F3NS/c1-3-8(4-2)9(5,6)7/h3-4H2,1-2H3

InChI key

CSJLBAMHHLJAAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Fluorinating agent: reaction with alcohols and carbonyl compounds, Review
  • Review on nucleophilic fluorination.
  • Catalyst for Friedel-Crafts allylation using tertiary cyclopropyl silyl ethers and the rearrangement of homoallylic alcohols to unsaturated aldehydes.
  • Early introduction of a fluoromethyl group stabilizes the epoxide during further manipulations in the synthesis of 26-fluoro-epothilone.
  • Fluorinating agent for a variety of compounds, including thioethers, alkenols, and cyanohydrins.
  • Reagent for gem difluorination of ketopipecolinic acids.

recommended

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Self-react. D - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective introduction of a trifluoromethyl group into an unsaturated system
Tetrahedron Letters, 32, 5963-5963 (1991)
Fluorination with diethylaminosulfur trifluoride and related aminofluorosulfuranes
M. Hudlicky
Org. React., 35, 513-513 (1998)
Po-Chiao Lin et al.
The Journal of organic chemistry, 74(11), 4041-4048 (2009-05-16)
When saccharides bearing a sulfur, selenium, or oxygen substituent at the anomeric center and an unprotected hydroxyl group either at C-4 or C-6 were subjected to fluorination with DAST in dichloromethane, a regioselective migration of the anomeric substituent to the
Synthesis of monodeoxyfluorinated, mucin-type oligosaccharide fragments.
R L Thomas et al.
Carbohydrate research, 165(2), c14-c16 (1987-08-01)
Reaction of tertiary cyclopropyl silyl ethers with diethylaminosulfur trifluoride. Part 2: The Friedel-Crafts allylation and cyclopropylation of electron-rich aromatic compounds
Kirihara, Masayuki, et al.
Tetrahedron Letters, 47, 3777-3777 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service