Skip to Content
Merck
All Photos(1)

Key Documents

310247

Sigma-Aldrich

1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose

98%

Synonym(s):

β-D-Mannopyranose 1,3,4,6-tetra-O-acetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$795.00

R$795.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
R$795.00

About This Item

Empirical Formula (Hill Notation):
C14H20O10
CAS Number:
Molecular Weight:
348.30
Beilstein:
96634
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

R$795.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

98%

optical activity

[α]19/D −68°, c = 0.7 in pyridine

mp

160-161 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@H](OC(C)=O)O[C@@H]([C@H]([C@@H]1OC(C)=O)OC(C)=O)COC(C)=O

InChI

1S/C14H20O10/c1-6(15)20-5-10-12(21-7(2)16)13(22-8(3)17)11(19)14(24-10)23-9(4)18/h10-14,19H,5H2,1-4H3/t10-,11+,12-,13-,14-/m1/s1

InChI key

SHBHJRVMGYVXKK-XVIXHAIJSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service