All Photos(2)
About This Item
Linear Formula:
(C2H5)2NCH2CH2NHCH3
CAS Number:
Molecular Weight:
130.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.4296 (lit.)
bp
157-160 °C (lit.)
density
0.805 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCN(CC)CCNC
InChI
1S/C7H18N2/c1-4-9(5-2)7-6-8-3/h8H,4-7H2,1-3H3
InChI key
MKDYQLJYEBWUIG-UHFFFAOYSA-N
General description
The Michael addition of N,N-Diethyl-N′-methylethylenediamine to nocathiacin was studied.
Application
N,N-Diethyl-N′-methylethylenediamine was used as a derivatization agent in the quantitative determination of isocyanates by capillary electrophoresis with tris(2,2′-bipyridyl)ruthenium(II) electrochemiluminescence (ECL).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
105.8 °F - closed cup
Flash Point(C)
41 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Haijuan Li et al.
Electrophoresis, 30(22), 3926-3931 (2009-11-04)
CE with tris(2,2'-bipyridyl) ruthenium(II) (Ru(bpy)(3) (2+)) electrochemiluminescence (ECL) detection for the quantitative determination of isocyanates was first reported. Hexamethylene diisocyanate (HDI) and hexyl isocyanate (HI) were used as the model analytes. Commercially available N,N-diethyl-N'-methylethylenediamine was used as the derivatization reagent.
Organic reactions in frozen water: Michael addition of amines and thiols to the dehydroalanine side chain of nocathiacins.
Tetrahedron Letters, 45(5), 1059-1063 null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service