Skip to Content
Merck
All Photos(1)

Key Documents

307912

Sigma-Aldrich

Boron trifluoride dibutyl etherate

Synonym(s):

Boron trifluoride butyl ether, Boron trifluoride butyl etherate, Dibutyl ether-boron trifluoride, Trifluoroboron dibutyl etherate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
R$1,265.00

R$1,265.00


Available to ship onApril 15, 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
R$1,265.00

About This Item

Linear Formula:
[CH3(CH2)3]2O·BF3
CAS Number:
Molecular Weight:
198.03
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

R$1,265.00


Available to ship onApril 15, 2025Details


Request a Bulk Order

form

liquid

Quality Level

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

density

0.959 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCCC[O+](CCCC)[B-](F)(F)F

InChI

1S/C8H18BF3O/c1-3-5-7-13(8-6-4-2)9(10,11)12/h3-8H2,1-2H3

InChI key

SLFVZPFCQQKCNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service