Skip to Content
Merck
All Photos(1)

Key Documents

306002

Sigma-Aldrich

3-Chloro-4-fluorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CHO
CAS Number:
Molecular Weight:
158.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

28-30 °C (lit.)

functional group

aldehyde
chloro
fluoro

SMILES string

Fc1ccc(C=O)cc1Cl

InChI

1S/C7H4ClFO/c8-6-3-5(4-10)1-2-7(6)9/h1-4H

InChI key

GVORVQPNNSASDM-UHFFFAOYSA-N

Application

3-Chloro-4-fluorobenzaldehyde was used in synthesis of triclosan analogs bearing isoxazole group on ring.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen P Muench et al.
Bioorganic & medicinal chemistry letters, 23(12), 3551-3555 (2013-05-15)
The enoyl acyl-carrier protein reductase (ENR) enzyme is harbored within the apicoplast of apicomplexan parasites providing a significant challenge for drug delivery, which may be overcome through the addition of transductive peptides, which facilitates crossing the apicoplast membranes. The binding
James P Hagen et al.
Bioorganic chemistry, 97, 103590-103590 (2020-03-18)
Insect-borne parasite Trypanosoma brucei plagues humans and other animals, eliciting the disease Human African trypanosomiasis, also known as African sleeping sickness. This disease poses the biggest threat to the people in Sub-Saharan Africa. Given the high toxicity and difficulties with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service