Skip to Content
Merck
All Photos(1)

Documents

305545

Sigma-Aldrich

N-Isopropyl-1,3-propanediamine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNH(CH2)3NH2
CAS Number:
Molecular Weight:
116.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.4425 (lit.)

bp

162 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

SMILES string

CC(C)NCCCN

InChI

1S/C6H16N2/c1-6(2)8-5-3-4-7/h6,8H,3-5,7H2,1-2H3

InChI key

KFDIDIIKNMZLRZ-UHFFFAOYSA-N

General description

Formation constant of complexes of copper, nickel, cobalt and zinc ions with N-isopropyl-1,3-propanediamine was evaluated.

Application

N-Isopropyl-1,3-propanediamine was used in preparation of cyclic urea.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Preparation of Polymeric and Cyclic Urethans and Ureas from Ethylene Carbonate and Amines.
Dyer E and Scott H.
Journal of the American Chemical Society, 79(3), 672-675 (1957)
Equilibrium Constants for the Formation of Complexes between Metal Ions and Polyamines1, 2.
Hares GB, et al.
Journal of the American Chemical Society, 78(9), 1816-1818 (1956)
Leslie S Hamachi et al.
Chemical science, 10(26), 6539-6552 (2019-08-02)
We report a method to control the composition and microstructure of CdSe1-x S x

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service