Skip to Content
Merck
All Photos(1)

Key Documents

302023

Sigma-Aldrich

(−)-2-Oxo-4-thiazolidinecarboxylic acid

97%

Synonym(s):

(R)-(−)-2-Oxothiazolidine-4-carboxylic acid, L-(−)-2-Oxothiazolidine-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NO3S
CAS Number:
Molecular Weight:
147.15
Beilstein:
4179169
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]19/D −60°, c = 1 in H2O

mp

174 °C (dec.) (lit.)

SMILES string

OC(=O)[C@@H]1CSC(=O)N1

Application

An intermediate in the total synthesis of (+)-latrunculin A.

replaced by

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 57, 5292-5292 (1992)
T-H Lee et al.
International endodontic journal, 43(5), 430-435 (2010-06-04)
To evaluate the mechanisms of cytotoxicity of chlorhexidine (CHX) in human osteoblastic cells in vitro. Cytotoxicity, cell proliferation and collagen synthesis assays were performed to elucidate the toxic effects of CHX on the human osteoblastic cell line U2OS. To determine
María José Clemente-Moreno et al.
Plant, cell & environment, 36(3), 640-654 (2012-08-24)
Sharka, a disease caused by plum pox virus (PPV), has a significant economic impact on fruit tree production. In this work, we analysed the effect of (2,1,3)-benzothiadiazole (BTH) and L-2-oxo-4-thiazolidine-carboxylic acid (OTC) on plant growth and virus content. OTC reduced
Sandra Hodge et al.
American journal of respiratory cell and molecular biology, 44(5), 673-681 (2010-07-03)
Defective efferocytosis may perpetuate inflammation in smokers with or without chronic obstructive pulmonary disease (COPD). Macrophages may phenotypically polarize to classically activated M1 (proinflammatory; regulation of antigen presentation) or alternatively activated M2 (poor antigen presentation; improved efferocytosis) markers. In bronchoalveolar
Anna Vávrová et al.
Archives of toxicology, 85(5), 525-535 (2010-11-04)
The clinical usefulness of anthracycline antineoplastic drugs is limited by their cardiotoxicity. Its mechanisms have not been fully understood, although the induction of oxidative stress is widely believed to play the principal role. Glutathione is the dominant cellular antioxidant, while

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service