Skip to Content
Merck
All Photos(2)

Key Documents

300632

Sigma-Aldrich

6,7-Dimethoxy-2,2-dimethyl-4-chromanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O4
CAS Number:
Molecular Weight:
236.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

105-107 °C (lit.)

functional group

ketone

SMILES string

COc1cc2OC(C)(C)CC(=O)c2cc1OC

InChI

1S/C13H16O4/c1-13(2)7-9(14)8-5-11(15-3)12(16-4)6-10(8)17-13/h5-6H,7H2,1-4H3

InChI key

OMWVNQFGCGNZHE-UHFFFAOYSA-N

Application

6,7-Dimethoxy-2,2-dimethyl-4-chromanone was used to examine the response of P. sojae zoospores to a wide range of compounds having some structural similarity to genistein and daidzein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B M Tyler et al.
Applied and environmental microbiology, 62(8), 2811-2817 (1996-08-01)
The zoospores of Phytophthora sojae are chemotactically attracted to the isoflavones genistein and daidzein that are released by soybean roots. In this study we have examined the response of P. sojae zoospores to a wide range of compounds having some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service