Skip to Content
Merck
All Photos(2)

Key Documents

298557

Sigma-Aldrich

7-(Carboxymethoxy)-4-methylcoumarin

97%

Synonym(s):

2-(4-Methyl-2-oxochromen-7-yl)oxyacetic acid, 2-[(4-Methyl-2-oxo-2H-1-benzopyran-7-yl)oxy]acetic acid, 2-[(4-Methyl-2-oxo-2H-chromen-7-yl)oxy]acetic acid, 7-Carboxymethoxy-4-methylchromen-2-one, [(4-Methyl-2-oxo-2H-chromen-7-yl)oxy]acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$254.00

R$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$254.00

About This Item

Empirical Formula (Hill Notation):
C12H10O5
CAS Number:
Molecular Weight:
234.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$254.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

206-208 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

CC1=CC(=O)Oc2cc(OCC(O)=O)ccc12

InChI

1S/C12H10O5/c1-7-4-12(15)17-10-5-8(2-3-9(7)10)16-6-11(13)14/h2-5H,6H2,1H3,(H,13,14)

InChI key

SGCOMUOACCXIKH-UHFFFAOYSA-N

General description

Synthesis of a novel fluorescent probe, Tb(III) - (7-carboxymethoxy-4-methylcoumarin) complex, for sensing of DNA has been reported.[1]

Application

7-(Carboxymethoxy)-4-methylcoumarin (CMC) was used in the preparation of poly(4-vinyl pyridine)-CMC, photosensitive polymer[2] and 3-chloro-7-[(chlorocarbonyl)methoxy]-4-methylcoumarin.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses of 3-Chloro-7-[(Chlorocarbonyl) Methoxy]-4-Methylcoumarin.
Phillips LR, et al.
Synthetic Communications, 26(9), 1805-1814 (1996)
Synthesis of novel fluorescent probe Tb (III)-7-carboxymethoxy-4-methylcoumarin complex for sensing of DNA.
Hussein BHM, et al.
Journal of Luminescence, 134, 441-446 (2013)
Photoinduced bending of a coumarin-containing supramolecular polymer.
He J, et al.
Soft Matter, 5(2), 308-310 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service