Skip to Content
Merck
All Photos(1)

Key Documents

297801

Sigma-Aldrich

Methyl 2-amino-3,5-dibromobenzoate

98%

Synonym(s):

Methyl 3,5-dibromoanthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H2-2-(NH2)CO2CH3
CAS Number:
Molecular Weight:
308.95
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

89-91 °C (lit.)

SMILES string

COC(=O)c1cc(Br)cc(Br)c1N

InChI

1S/C8H7Br2NO2/c1-13-8(12)5-2-4(9)3-6(10)7(5)11/h2-3H,11H2,1H3

InChI key

NGXVMFCGYYHEGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 2-amino-3,5-dibromobenzoate participates in cobalt(II) Schiff′s base complex catalyzed oxidation of primary and secondary alcohols to aldehydes and ketones respectively[1]. It also participates in allylic and benzylic oxidation of various cyclic alkenes and benzylic compounds respectively[2].

Application

Methyl 2-amino-3,5-dibromobenzoate was used in the preparation of ethyl 1-(p-methoxyphenyl)-2-oxocyclopentanecarboxylate via reaction with chloroform containing pyridine[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cobalt catalysed allylic and benzylic oxidations with dioxygen in the presence of ethyl 2-oxocyclopentanecarboxylate.
Punniyamurthy T and Iqbal J.
Tetrahedron Letters, 35(23), 4003-4006 (1994)
The chemistry of aryllead (IV) tricarboxylates. Reaction with ?-keto esters: a convenient route to a-arylated ketones.
Pinhey JT and Rowe BA.
Australian Journal of Chemistry, 33(1), 113-120 (1980)
Cobalt (II) Schiff's base complex catalysed oxidation of alcohols with dioxygen in the presence of ethyl 2-oxocyclopentanecarboxylate.
Punniyamurthy T and Iqbal J.
Tetrahedron Letters, 35(23), 4007-4010 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service