Skip to Content
Merck
All Photos(1)

Key Documents

292869

Sigma-Aldrich

2-Aminoethyl dihydrogen phosphate

98%

Synonym(s):

O-Phosphocolamine, O-Phosphoethanolamine, O-Phosphorylethanolamine, Colamine phosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)2OP(O)(OH)2
CAS Number:
Molecular Weight:
141.06
Beilstein:
1758916
EC Number:
MDL number:
UNSPSC Code:
12161700

Assay

98%

mp

241-243 °C (lit.)

SMILES string

NCCOP(O)(O)=O

InChI

1S/C2H8NO4P/c3-1-2-7-8(4,5)6/h1-3H2,(H2,4,5,6)

InChI key

SUHOOTKUPISOBE-UHFFFAOYSA-N

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret N Holme et al.
Nature nanotechnology, 7(8), 536-543 (2012-06-12)
Atherosclerosis results in the narrowing of arterial blood vessels and this causes significant changes in the endogenous shear stress between healthy and constricted arteries. Nanocontainers that can release drugs locally with such rheological changes can be very useful. Here, we
Lin Zhu et al.
ACS nano, 6(4), 3491-3498 (2012-03-14)
A novel "smart" multifunctional drug delivery system was successfully developed to respond to the up-regulated matrix metalloprotease 2 (MMP2) in the tumor microenvironment and improve cancer cell-specific delivery of loaded drugs. The system represents a surface-functionalized liposomal nanocarrier, for which
Christopher Wanty et al.
Journal of molecular biology, 425(18), 3389-3402 (2013-07-03)
Gram-negative bacteria possess an outer membrane envelope consisting of an outer leaflet of lipopolysaccharides, also called endotoxins, which protect the pathogen from antimicrobial peptides and have multifaceted roles in virulence. Lipopolysaccharide consists of a glycan moiety attached to lipid A
Vanessa Gonzalez-Covarrubias et al.
Aging cell, 12(3), 426-434 (2013-03-05)
Middle-aged offspring of nonagenarians, as compared to their spouses (controls), show a favorable lipid metabolism marked by larger LDL particle size in men and lower total triglyceride levels in women. To investigate which specific lipids associate with familial longevity, we
Conan K Wang et al.
The Journal of biological chemistry, 287(52), 43884-43898 (2012-11-07)
Cyclotides are a family of plant-derived circular proteins with potential therapeutic applications arising from their remarkable stability, broad sequence diversity, and range of bioactivities. Their membrane-binding activity is believed to be a critical component of their mechanism of action. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service