Skip to Content
Merck
All Photos(2)

Documents

292508

Sigma-Aldrich

2,6-Dimethoxybenzaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CHO
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

285 °C (lit.)

mp

96-98 °C (lit.)

SMILES string

COc1cccc(OC)c1C=O

InChI

1S/C9H10O3/c1-11-8-4-3-5-9(12-2)7(8)6-10/h3-6H,1-2H3

InChI key

WXSGQHKHUYTJNB-UHFFFAOYSA-N

General description

Demethylation of 2,6-dimethoxybenzaldehydes with magnesium iodide etherate has been studied.

Application

2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective demethylation of 2, 6-dimethoxybenzaldehydes with magnesium iodide etherate.
Yamaguchi S, et al.
Tetrahedron Letters, 40(41), 7363-7365 (1999)
Synthesis of facially-encumbered porphyrins. An approach to light-harvesting antenna complexes.
Wagner RW, et al.
Tetrahedron Letters, 32(14), 1703-1706 (1991)
Olga V Kupriyanova et al.
Drug testing and analysis, 12(8), 1154-1170 (2020-05-18)
N-(2-Methoxybenzyl)-2,5-dimethoxyphenethylamines (NBOMes) are synthetic phenethylamine derivatives emerging on the global drug market and reported to be associated with untoward effects in people who use drugs. Its action involves agonism at serotonin 5-HT2A receptors, affecting cognitive and behavioral processes. However, certain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service