Skip to Content
Merck
All Photos(1)

Documents

292214

Sigma-Aldrich

3-Acetylbenzonitrile

97%

Synonym(s):

3′-Cyanoacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC6H4CN
CAS Number:
Molecular Weight:
145.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

98-100 °C (lit.)

SMILES string

CC(=O)c1cccc(c1)C#N

InChI

1S/C9H7NO/c1-7(11)9-4-2-3-8(5-9)6-10/h2-5H,1H3

InChI key

SBCFGFDAZCTSRH-UHFFFAOYSA-N

Application

3-Acetylbenzonitrile has been used in the preparation of:
  • 3-(hydroxyacetyl)benzonitrile
  • 3-(1-(3-(4-(pyridin-3-yl)pyrimidin-2-ylamino)-4-methylphenylimino)ethyl)benzonitrile

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and in vitro biological activity of N-(5-amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidinamine derivatives.
Mohana KN and Mallesha L.
Bulgarian Chemical Communications, 43(3), 395-400 (2011)
C Y Watson et al.
Bioorganic & medicinal chemistry, 6(6), 721-734 (1998-07-29)
Inhibitors of poly(ADP-ribose)polymerase (PARP) inhibit repair of damaged DNA and thus potentiate radiotherapy and chemotherapy of cancer. 3-Substituted benzamides and 5-substituted isoquinolin-1-ones have been synthesised and evaluated for inhibition of PARP. Reduction of 3-(bromoacetyl)benzamide, followed by treatment with base, gave

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service