291552
6-Methoxy-1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole
97%
Synonym(s):
6-Methoxy-1,2,3,4-tetrahydro-β-carboline
About This Item
Recommended Products
Assay
97%
form
powder or crystals
mp
219-222 °C (lit.)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
COc1ccc2[nH]c3CNCCc3c2c1
InChI
1S/C12H14N2O/c1-15-8-2-3-11-10(6-8)9-4-5-13-7-12(9)14-11/h2-3,6,13-14H,4-5,7H2,1H3
InChI key
QYMDEOQLJUUNOF-UHFFFAOYSA-N
Gene Information
rat ... Htr2a(29595) , Htr2c(25187)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service