Skip to Content
Merck
All Photos(2)

Key Documents

289078

Sigma-Aldrich

Gold′s Reagent

95%

Synonym(s):

(Dimethylaminomethyleneaminomethylene)dimethylammonium chloride, [3-(Dimethylamino)-2-azaprop-2-en-1-ylidene]dimethylammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$640.00

R$640.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

Select a Size

Change View
25 G
R$640.00

About This Item

Linear Formula:
(CH3)2NCH=NCH=N(CH3)2Cl
CAS Number:
Molecular Weight:
165.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$640.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

Assay

95%

form

solid

mp

100-102 °C (lit.)

functional group

amine

SMILES string

[Cl-].CN(C)\C=N\C=[N+](\C)C

InChI

1S/C6H14N3.ClH/c1-8(2)5-7-6-9(3)4;/h5-6H,1-4H3;1H/q+1;/p-1

InChI key

DEIBXAPEZDJDRC-UHFFFAOYSA-M

Application

Gold′s Reagent was used in the synthesis of 4-hydroxyquinazoline by reacting with ortho aminobenzoic acids[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gupton JT, et al.
Synthetic Communications, 14(11), 1013-1025 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service