Skip to Content
Merck
All Photos(1)

Key Documents

284858

Sigma-Aldrich

(−)-Norgestrel

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −32°, c = 1 in chloroform

mp

239-241 °C (lit.)

SMILES string

[H][C@]12CCC(=O)C=C1CC[C@]3([H])[C@]2([H])CC[C@@]4(CC)[C@@]3([H])CC[C@@]4(O)C#C

InChI

1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1

InChI key

WWYNJERNGUHSAO-XUDSTZEESA-N

Gene Information

human ... PGR(5241)
rat ... Ar(24208)

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice W Y Wong et al.
Obstetrics and gynecology, 121(5), 943-950 (2013-05-03)
To estimate the rate of endometrial pathology with the prophylactic use of levonorgestrel-releasing intrauterine system in women with breast cancer treated with tamoxifen. This was a randomized contro-lled trial of 129 Chinese women who attended a university hospital in Hong
Satu Heliövaara-Peippo et al.
American journal of obstetrics and gynecology, 209(6), 535-535 (2013-09-04)
Menorrhagia is a common problem impairing the quality of life (QOL) of many women. Both levonorgestrel-releasing intrauterine system (LNG-IUS) and hysterectomy are effective treatment modalities but no long-term comparative studies of QOL and costs exist. The objective of this study
Elizabeth A McCarthy et al.
Contraception, 86(5), 587-590 (2012-04-14)
Correctly placed levonorgestrel-releasing intrauterine systems (LNG-IUSs) are rarely associated with intrauterine pregnancy when pregnancy occurs. LNG-IUS retrieval, termination of pregnancy and conservative management if retracted strings prevent ready removal are the usual clinical options given to women. The conservative course
Abbey B Berenson et al.
Obstetrics and gynecology, 121(5), 951-958 (2013-05-03)
Many U.S. health care providers remain reluctant to prescribe intrauterine devices (IUDs) to teenagers as a result of concerns about serious complications. This study examined whether 15-19-year-old IUD users were more likely to experience complications, failure, or early discontinuation than
Anita Nelson et al.
Obstetrics and gynecology, 122(6), 1205-1213 (2013-11-19)
To evaluate the efficacy and safety of two low-dose levonorgestrel intrauterine contraceptive systems. Nulliparous and parous women aged 18-35 years with regular menstrual cycles (21-35 days) requesting contraception were randomized to 3 years of treatment with one of two levonorgestrel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service