Skip to Content
Merck
All Photos(1)

Key Documents

283363

Sigma-Aldrich

3-Fluoro-4-nitrotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)F
CAS Number:
Molecular Weight:
155.13
Beilstein:
2502584
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

bp

97-98 °C/3 mmHg (lit.)

mp

52-55 °C (lit.)

functional group

fluoro
nitro

SMILES string

Cc1ccc(c(F)c1)[N+]([O-])=O

InChI

1S/C7H6FNO2/c1-5-2-3-7(9(10)11)6(8)4-5/h2-4H,1H3

InChI key

WZMOWQCNPFDWPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Fluoro-4-nitrotoluene has been used in the preparation of IκB kinase (IKK) inhibitor, BMS [N1-(1,8-dimethylimidazo[1,2-a]quinolin-4-yl)-ethane-1,2-diamine].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoko Hayashi et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(47), 18619-18624 (2007-11-16)
3-Hydroxyanthranilic acid (HAA), a compound generated during tryptophan metabolism initiated by indoleamine 2,3-dioxygenase, is known to induce T cell death, but its molecular target is not known. Here we report that HAA inhibits NF-kappaB activation upon T cell antigen receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service