Skip to Content
Merck
All Photos(2)

Key Documents

278890

Sigma-Aldrich

Iodopentoxide

≥98%

Synonym(s):

Diiodine pentoxide, Iodic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$976.00
100 G
R$3,074.00

R$976.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
R$976.00
100 G
R$3,074.00

About This Item

Empirical Formula (Hill Notation):
I2O5
CAS Number:
Molecular Weight:
333.81
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

R$976.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98%

density

5.08 g/mL at 25 °C

SMILES string

O=I(=O)OI(=O)=O

InChI

1S/I2O5/c3-1(4)7-2(5)6

InChI key

BIZCJSDBWZTASZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

I2O5 can be used as a metal free catalyst for the difunctionalization of indoles with thiophenols.

It can be for the dibromination and bromohydroxylation of olefins.

Passivated iodopentoxide can be used as an iodine-richoxidizer in nanoenergetic formulations.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service