Skip to Content
Merck
All Photos(1)

Key Documents

27820

Sigma-Aldrich

Poly(styrene-co-divinylbenzene)

99% styrene – 1% DVB

Synonym(s):

Polystyrene crosslinked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH2CH[C6H4(CHCH2)]]y
CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

particle size

200-400 mesh

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C=Cc3cccc(C=C)c3.C=Cc4ccccc4C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of Chromosorb 102 to air samples pre-concentration in the determination of low concentrated dibutylphthalate.
L Wolska
Bulletin of the Institute of Maritime and Tropical Medicine in Gdynia, 39(1-2), 81-89 (1988-01-01)
E G Spack et al.
Analytical biochemistry, 158(1), 233-237 (1986-10-01)
Rhodamine absorbed to protein was removed from rhodamine-conjugated antibody solutions by adsorption to hydrophobic macroporous beads (Bio-Beads SM-2) following gel permeation chromatography. This simple technique eliminated the contaminating free dye more effectively than gel filtration alone, but neither significantly reduced
Sibel Saracoglu et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 19(2), 259-264 (2003-03-01)
The selective retention of the Sb(III) chelate with ammonium pyrrolidine dithiocarbamate (APDC) on a column of Chromosorb 102 resin from a buffered sample solution including Sb(V) was used for the determination of Sb(III). The retained antimony was eluted with acetone.
T C Thomas et al.
American Industrial Hygiene Association journal, 41(8), 599-601 (1980-08-01)
An organic polymeric solid, Chromosorb 102, has been reported to collect various pesticides from air. In this study the collection efficiency of Chromosorb 102 for chlordane was evaluated. Preparation of sampling tubes containing this medium for field sampling is also
Sampling of airborne pesticides using Chromosorb 102.
T C Thomas et al.
Bulletin of environmental contamination and toxicology, 35(4), 460-465 (1985-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service