Skip to Content
Merck
All Photos(1)

Key Documents

276111

Sigma-Aldrich

Diethyl nitromalonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NCH(CO2C2H5)2
CAS Number:
Molecular Weight:
205.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

78-80 °C/0.2 mmHg (lit.)

density

1.174 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C(=O)OCC)[N+]([O-])=O

InChI

1S/C7H11NO6/c1-3-13-6(9)5(8(11)12)7(10)14-4-2/h5H,3-4H2,1-2H3

InChI key

DAKKWKYIQGMKOS-UHFFFAOYSA-N

General description

Thermal decomposition of diethyl nitromalonate to furoxan has been studied.

Application

Diethyl nitromalonate has been used in the preparation of 5,5-diethoxycarbonyl-1-pyrroline N-oxide (DECPO).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structure of 5, 5-diethoxycarbonyl-1-pyrroline< i> N</i>-oxide (DECPO). Application to superoxide radical trapping.
Karoui H, et al.
Tetrahedron Letters, 45(1), 149-152 (2004)
A New Synthetic Method of Alkyl Carbonocyanidate iV-Oxides.
SHiMizu T, et al.
Bulletin of the Chemical Society of Japan, 58, 2519-2522 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service