Skip to Content
Merck
All Photos(1)

Documents

273112

Sigma-Aldrich

(1R,E)-(+)-Camphorquinone 3-oxime

99%

Synonym(s):

(1R,E)-(+)-2,3-Bornanedione 3-oxime, anti-(1R)-(+)-Camphorquinone 3-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO2
CAS Number:
Molecular Weight:
181.23
Beilstein:
3200377
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]25/D +200°, c = 1 in ethanol

mp

153-156 °C (lit.)

SMILES string

[H][C@@]12CC[C@@](C)(C(=O)\C1=N/O)C2(C)C

InChI

1S/C10H15NO2/c1-9(2)6-4-5-10(9,3)8(12)7(6)11-13/h6,13H,4-5H2,1-3H3/b11-7-/t6-,10+/m1/s1

InChI key

YRNPDSREMSMKIY-HAKKTOSXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(1R,E)-(+)-Camphorquinone 3-oxime can be used to prepare chiral camphor-oxazoline auxiliary, which is used to blend diastereoselectivity and isomer separability for the efficient synthesis of bicuculline, egenine, corlumine, and corytensine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-Magnesiotetrahydroisoquinolyloxazolines as Chiral Nucleophiles in Stereoselective Additions to Aldehydes: Auxiliary Optimization, Asymmetric Synthesis of (+)-Corlumine,(+)-Bicuculline,(+)-Egenine, and (+)-Corytensine, and Preliminary 13C NMR Studies of 1-Lithio-and 1-Magnesiotetrahydroisoquinolyloxazolines
Gawley RE and Z Pingsheng
The Journal of Organic Chemistry, 61(23), 8103- 8112 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service