Skip to Content
Merck
All Photos(1)

Documents

261327

Sigma-Aldrich

Yttrium

powder, −40 mesh, 99.5% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
EC Number:
MDL number:
UNSPSC Code:
12141617
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5% trace rare earth metals basis

form

powder

resistivity

57 μΩ-cm, 20°C

particle size

−40 mesh

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Blanco-Canosa et al.
Physical review letters, 110(18), 187001-187001 (2013-05-21)
We use resonant x-ray scattering to determine the momentum-dependent charge correlations in YBa(2)Cu(3) O(6.55) samples with highly ordered chain arrays of oxygen acceptors (ortho-II structure). The results reveal nearly critical, biaxial charge density wave (CDW) correlations at in-plane wave vectors
Yttrium-catalyzed addition of benzylic C-H bonds of alkyl pyridines to olefins.
Bing-Tao Guan et al.
Angewandte Chemie (International ed. in English), 52(16), 4418-4421 (2013-03-21)
José Renato Queiroz et al.
The journal of adhesive dentistry, 15(2), 151-159 (2013-03-28)
To compare the effect of silica (Si)-based nano-coating deposited by reactive magnetron sputtering (RMP) with that of conventional surface conditioning using metal/zirconia primer alone or after air-particle abrasion on the adhesion of resin cements to zirconia ceramic. Two hundred forty
Masahiko Ooe et al.
Aesthetic plastic surgery, 37(2), 424-433 (2013-02-12)
Skin wrinkles are one of the most cosmetically concerning signs of aging for women, and improvements in the visual effect of wrinkles become a matter of concern that has an impact on the quality of life. Although various wrinkle treatments
Gitanjal Deka et al.
Journal of biomedical optics, 19(1), 011012-011012 (2013-08-21)
Cellular micropattering has been increasingly adopted in quantitative biological experiments. A Q-switched pulsed neodymium-doped yttrium ortho-vanadate (Nd∶YVO4) laser directed in-situ microfabrication technique for cell patterning is presented. A platform is designed uniquely to achieve laser ablation. The platform is comprised

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service