Skip to Content
Merck
All Photos(2)

Key Documents

259500

Sigma-Aldrich

1-Nitropentane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$652.00

R$652.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$652.00

About This Item

Linear Formula:
CH3(CH2)4NO2
CAS Number:
Molecular Weight:
117.15
Beilstein:
506508
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$652.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

75-76 °C/23 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)

functional group

amine
nitro

SMILES string

CCCCC[N+]([O-])=O

InChI

1S/C5H11NO2/c1-2-3-4-5-6(7)8/h2-5H2,1H3

InChI key

BVALZCVRLDMXOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ferroxime(II)-catalyzed oxidative denitrification of 1-nitropentane has been reported[1].

Application

1-Nitropentane has been used as organic nitrogen (ON) standard in quantitative method for the speciation of ON within ambient atmospheric aerosol[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferroxime (II)-catalyzed oxygenation of nitroalkanes by tert-butyl hydroperoxide and its relevance to 2-nitropropane dioxygenase.
Kaizer J, et al.
J. Mol. Catal. A: Chem., 179(1), 35-39 (2002)
Mustafa Z Özel et al.
Environmental science & technology, 45(4), 1497-1505 (2011-01-08)
Atmospheric aerosols contain a highly complex mixture of organic and inorganic compounds; however, as a chemical class relatively little is known about organic nitrogen (ON) content, with few satisfactory methods for speciated analysis. In this paper we report a sensitive

Articles

A shortened review of fundamentals of Porous Graphitic Carbon (PGC) materials. What they are and how do they work as a stationary phase material in HPLC?.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service