Skip to Content
Merck
All Photos(2)

Key Documents

258717

Sigma-Aldrich

3,4-Dimethoxy-5-hydroxybenzaldehyde

98%

Synonym(s):

5-Hydroxyveratraldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$449.00

R$449.00


Estimated to ship onMay 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
R$449.00

About This Item

Linear Formula:
HOC6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$449.00


Estimated to ship onMay 01, 2025


Request a Bulk Order

Quality Level

Assay

98%

form

powder

bp

177-180 °C/12 mmHg (lit.)

mp

67-69 °C (lit.)

functional group

aldehyde

SMILES string

COc1cc(C=O)cc(O)c1OC

InChI

1S/C9H10O4/c1-12-8-4-6(5-10)3-7(11)9(8)13-2/h3-5,11H,1-2H3

InChI key

NVLTWXMZECWWPC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy J Tschaplinski et al.
Biotechnology for biofuels, 5(1), 71-71 (2012-09-25)
Down-regulation of the caffeic acid 3-O-methyltransferase EC 2.1.1.68 (COMT) gene in the lignin biosynthetic pathway of switchgrass (Panicum virgatum) resulted in cell walls of transgenic plants releasing more constituent sugars after pretreatment by dilute acid and treatment with glycosyl hydrolases
Carsten Höltke et al.
Bioorganic & medicinal chemistry, 17(20), 7197-7208 (2009-09-22)
The expression and function of endothelin (ET) receptors is abnormal in cardiovascular diseases, tumor progression, and tumor metastasis. In this study, we prepared two [(18)F]-fluorinated derivatives of the non-peptide ET(A) receptor antagonist PD 156707 and evaluated their ET receptor binding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service