Skip to Content
Merck
All Photos(1)

Key Documents

258377

Sigma-Aldrich

Protoporphyrin IX dimethyl ester

from ox hemin

Synonym(s):

Dimethyl 8,13-divinyl-3,7,12,17-tetramethyl-21H,23H-porphine-2,18-dipropionate, Ooporpyhrin dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H38N4O4
CAS Number:
Molecular Weight:
590.71
Beilstein:
381574
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

225-228 °C (lit.)

SMILES string

COC(=O)CCc1c(C)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(CCC(=O)OC)c5C)c(C)c4C=C)c(C)c3C=C

InChI

1S/C36H38N4O4/c1-9-23-19(3)27-15-28-21(5)25(11-13-35(41)43-7)33(39-28)18-34-26(12-14-36(42)44-8)22(6)30(40-34)17-32-24(10-2)20(4)29(38-32)16-31(23)37-27/h9-10,15-18,38-39H,1-2,11-14H2,3-8H3/b27-15-,28-15-,29-16-,30-17-,31-16-,32-17-,33-18-,34-18-

InChI key

XNCGCBXDDMJCKW-MFBGAUBSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Protoporphyrin IX dimethyl ester has been demonstrated to a potent photosensitizer of human nasopharyngeal carcinoma.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service