255831
Germanium(IV) chloride
99.999% trace metals basis
Synonym(s):
Germanium tetrachloride
About This Item
Recommended Products
vapor pressure
76 mmHg ( 20 °C)
Assay
99.999% trace metals basis
form
liquid
reaction suitability
reagent type: catalyst
core: germanium
impurities
≤15.0 ppm Trace Metal Analysis
bp
83 °C (lit.)
mp
−49.5 °C (lit.)
density
1.88 g/mL at 25 °C
SMILES string
Cl[Ge](Cl)(Cl)Cl
InChI
1S/Cl4Ge/c1-5(2,3)4
InChI key
IEXRMSFAVATTJX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Does not sustain combustion.
Flash Point(C)
Does not sustain combustion.
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service