Skip to Content
Merck
All Photos(1)

Documents

252255

Sigma-Aldrich

2-Chlorobenzyl bromide

97%

Synonym(s):

α-Bromo-2-chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2Br
CAS Number:
Molecular Weight:
205.48
Beilstein:
386264
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.592 (lit.)

bp

103-104 °C/10 mmHg (lit.)

density

1.583 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1CBr

InChI

1S/C7H6BrCl/c8-5-6-3-1-2-4-7(6)9/h1-4H,5H2

InChI key

PURSZYWBIQIANP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chlorobenzyl bromide participates in Hass-Bender oxidation as well as the Henry reaction.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tandem Hass-Bender/Henry reaction for the synthesis of dimethylnitro alcohols from benzylic halides.
Klein TA and Schkeryantz JM.
Tetrahedron Letters, 46(27), 4535-4538 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service